missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,5-Dichlorosulfanilate 99.5+%, TCI America™

Catalog No. D041625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D041625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D041625G Supplier TCI America Supplier No. D041625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41295-98-1
Molecular Formula C6H4Cl2NNaO3S
Molecular Weight (g/mol) 264.052
MDL Number MFCD00068356
InChI Key MOJYHTGMAPQZMC-UHFFFAOYSA-M
Synonym 2,5-Dichloroaniline-4-sulfonic Acid Sodium Salt, Sodium 2,5-Dichloroaniline-4-sulfonate, 2,5-Dichlorosulfanilic Acid Sodium Salt
PubChem CID 23688979
IUPAC Name sodium;4-amino-2,5-dichlorobenzenesulfonate
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)[O-])Cl)N.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 264.05
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,5-Dichlorosulfanilate
EINECSNumber (3)-2086
TSCA Yes