missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,2-Dichloropropionate 85.0+%, TCI America™

Catalog No. D040925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D040925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D040925G Supplier TCI America Supplier No. D040925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127-20-8
Molecular Formula C3H3Cl2NaO2
Molecular Weight (g/mol) 164.945
MDL Number MFCD00064199
InChI Key PDEFQWNXOUGDJR-UHFFFAOYSA-M
Synonym 2,2-Dichloropropionic Acid Sodium Salt
PubChem CID 517058
IUPAC Name sodium;2,2-dichloropropanoate
SMILES CC(C(=O)[O-])(Cl)Cl.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 164.94
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,2-Dichloropropionate
EINECSNumber (2)-2654
RTECSNumber UF1225000
TSCA Yes