missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%

Catalog No. aab2033222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2033222 100 g
AAB2033214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2033222 Supplier Thermo Scientific Chemicals Supplier No. B2033222
Only null left

CAS: 5838-34-6 | C20H14N4O4S | 406.42 g/mol

It can be used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as catalyst in pharmaceutical research.

Solubility
Solubility in water is almost transparent.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5838-34-6
Molecular Formula C20H14N4O4S
Molecular Weight (g/mol) 406.42
MDL Number MFCD00053627
InChI Key RFNALTSHRWGOHV-UHFFFAOYSA-N
Synonym sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1
PubChem CID 23692564
IUPAC Name sodium;undecane-1-sulfonate
SMILES O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 3751377
Formula Weight 258.36
Percent Purity ≥98%
Assay (dry wt.), water <2%
Chemical Name or Material Sodium 1-undecanesulfonate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only