missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-pentanesulfonate monohydrate, HPLC grade

Catalog No. p-7049598
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1571406 5 g
AA1571414 25 g
AA1571422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1571406 Supplier Thermo Scientific Chemicals Supplier No. 01571406

Ion pair reagent | CAS: 207605-40-1 | C5H13NaO4S | 192.205 g/mol

Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H13NaO4S
Molecular Weight (g/mol) 192.205
MDL Number MFCD00007541
InChI Key FPQYXAFKHLSWTI-UHFFFAOYSA-M
Synonym sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
PubChem CID 23693099
IUPAC Name sodium;pentane-1-sulfonate;hydrate
SMILES CCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Color White
Linear Formula CH3(CH2)4SO3Na·H2O
Quantity 5 g
Beilstein 3725147
Solubility Information Soluble in water.
Formula Weight 192.21 (174.19 Anhydrous)
Grade HPLC
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-pentanesulfonate monohydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-208-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only