missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-pentanesulfonate, 99% (dry wt.), water <1.5%

Catalog No. AAB2069922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2069922 100 g
AAB2069906 5 g
AAB2069914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2069922 Supplier Thermo Scientific Chemicals Supplier No. B2069922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol

Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium;pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 3725147
Solubility Information Soluble in water.
Formula Weight 174.19
Percent Purity 99%
Assay (dry wt.), water <1%
Chemical Name or Material Sodium 1-pentanesulfonate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-208-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only