missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%

Catalog No. p-7052135
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4263606 5 g
AA4263614 25 g
AA4263622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4263606 Supplier Thermo Scientific Chemicals Supplier No. 04263606

Ion-pair reagent for HPLC analysis | CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol

Ion-pair reagent for HPLC analysisSodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium;octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Soluble in water producing a clear and colorless solution.
Formula Weight 216.28 (Anhydrous)
Percent Purity ≥99%
Grade HPLC
Physical Form Crystalline
Chemical Name or Material Sodium 1-octanesulfonate hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-195-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only