missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-octanesulfonate, 99%

CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol

Supplier:  Thermo Scientific Chemicals A1429214

 View more versions of this product

Catalog No. AAA1429214


Only null left
Explore available promotions

Description

Description

Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium-1-Octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-Octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.

Solubility
Soluble in water producing a clear and colorless solution

Notes
Sodium D-gluconate is stable under normal temperatures.
Specifications

Specifications

Sodium 1-Octanesulfonate
>300°C
C8H17NaO3S
25 g
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium;octane-1-sulfonate
23669624
5324-84-5
99%
MFCD00007544
3574241
Soluble in water producing a clear and colorless solution
[Na+].CCCCCCCCS([O-])(=O)=O
216.27
216.28

Chemical Identifiers

5324-84-5
216.27
HRQDCDQDOPSGBR-UHFFFAOYSA-M
23669624
[Na+].CCCCCCCCS([O-])(=O)=O
C8H17NaO3S
MFCD00007544
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
sodium;octane-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-195-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only