missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-Nonanesulfonate 98.0+%, TCI America™
$92.41 - $358.08
Chemical Identifiers
| CAS | 35192-74-6 |
|---|---|
| Molecular Formula | C9H19NaO3S |
| Molecular Weight (g/mol) | 230.298 |
| MDL Number | MFCD00025010 |
| InChI Key | RUYRDULZOKULPK-UHFFFAOYSA-M |
| Synonym | sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate |
| PubChem CID | 23664777 |
| IUPAC Name | sodium;nonane-1-sulfonate |
| SMILES | CCCCCCCCCS(=O)(=O)[O-].[Na+] |
Chemical Identifiers
| 35192-74-6 | |
| 230.298 | |
| RUYRDULZOKULPK-UHFFFAOYSA-M | |
| 23664777 | |
| CCCCCCCCCS(=O)(=O)[O-].[Na+] |
| C9H19NaO3S | |
| MFCD00025010 | |
| sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate | |
| sodium;nonane-1-sulfonate |
Specifications
| 35192-74-6 | |
| C9H19NaO3S | |
| 5 g | |
| RUYRDULZOKULPK-UHFFFAOYSA-M | |
| sodium;nonane-1-sulfonate | |
| 23664777 | |
| ≥98.0% (T) | |
| Sodium 1-Nonanesulfonate [Reagent for Ion-Pair Chromatography] |
| White | |
| MFCD00025010 | |
| sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate | |
| CCCCCCCCCS(=O)(=O)[O-].[Na+] | |
| 230.298 | |
| 230.30 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1640
TSCA : No