missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Nonanesulfonate 98.0+%, TCI America™

Catalog No. I03475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03475G 5 g
I034725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03475G Supplier TCI America Supplier No. I03475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35192-74-6
Molecular Formula C9H19NaO3S
Molecular Weight (g/mol) 230.298
MDL Number MFCD00025010
InChI Key RUYRDULZOKULPK-UHFFFAOYSA-M
Synonym sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate
PubChem CID 23664777
IUPAC Name sodium;nonane-1-sulfonate
SMILES CCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 230.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Nonanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-1640
TSCA No