missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-hexadecanesulfonate, 97%

Catalog No. p-7025062
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2038206 5 g
AAB2038214 25 g
AAB2038222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2038206 Supplier Thermo Scientific Chemicals Supplier No. B2038206
Only null left

CAS: 15015-81-3 | C16H33NaO3S | 328.487 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15015-81-3
Molecular Formula C16H33NaO3S
Molecular Weight (g/mol) 328.487
MDL Number MFCD00007528
InChI Key PNGBYKXZVCIZRN-UHFFFAOYSA-M
Synonym sodium cetylsulfonate, sodium 1-hexadecanesulfonate, 1-hexadecanesulfonic acid sodium salt, 1-hexadecanesulfonic acid, sodium salt, sodium hexadecane-1-sulfonate, unii-sk4e885p15, sodium hexadecane-1-sulphonate, cetylsulfonic acid sodium salt, 1-hexadecanesulfonic acid, sodium salt 1:1, hexadecylsulfonic acid sodium salt
PubChem CID 23665769
IUPAC Name sodium;hexadecane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Linear Formula CH3(CH2)15SO3Na
Quantity 5 g
Beilstein 3920654
Formula Weight 328.49
Percent Purity 97%
Chemical Name or Material Sodium 1-hexadecanesulfonate
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-100-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only