missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Heptanesulfonate 98.0+%, TCI America™

Catalog No. p-7130544
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H045425G 25 g
H0454100G 100 g
H0454500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H045425G Supplier TCI America Supplier No. H045425G
Only null left

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number MFCD00007543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 202.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Heptanesulfonate