missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Heptanesulfonate 98.0+%, TCI America™

Catalog No. p-7131352
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
I03455G 5 g
I034525G 25 g
I0345100G 100 g
3 options

Catalog No. I03455G

Supplier: TCI America I03455G

Only null left

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number MFCD00007543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White
Quantity 5 g
Formula Weight 202.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Heptanesulfonate [Reagent for Ion-Pair Chromatography]