missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-heptanesulfonate monohydrate, 99%

Catalog No. p-7049583
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1521409 10 g
AA1521418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1521409 Supplier Thermo Scientific Chemicals Supplier No. 01521409

CAS: 207300-90-1 | C7H17NaO4S | 220.259 g/mol

Sodium 1-heptanesulfonate monohydrate is an ion pairing reagent used in the HPLC analysis of proteins and peptides. It is utilized in high performance capillary electrophoresis analysis of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207300-90-1
Molecular Formula C7H17NaO4S
Molecular Weight (g/mol) 220.259
MDL Number MFCD00149550
InChI Key XWZCREJRXRKIRQ-UHFFFAOYSA-M
Synonym sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
PubChem CID 23687711
IUPAC Name sodium;heptane-1-sulfonate;hydrate
SMILES CCCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Melting Point >300°C
Color White
Linear Formula CH3(CH2)6SO3Na·H2O
Quantity 10 g
Beilstein 3731762
Solubility Information Slightly soluble in water.
Formula Weight 220.26 (202.24 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Sodium 1-heptanesulfonate monohydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-210-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only