missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%

Catalog No. AAA1091706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1091706 5 g
AAA1091714 25 g
AAA1091722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1091706 Supplier Thermo Scientific Chemicals Supplier No. A1091706

CAS: 22767-50-6 | C7H15NaO3S | 202.244 g/mol

HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number MFCD00007543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Beilstein 3731762
Solubility Information Soluble in water.
Formula Weight 202.24
Percent Purity 99%
Assay (dry wt.)
Chemical Name or Material Sodium 1-heptanesulfonate, Water <2%
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-210-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only