missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Decanesulfonate 98.0+%, TCI America™

Catalog No. p-7126629
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D11815G 5 g
D118125G 25 g
2 options

Catalog No. D11815G

Supplier: TCI America D11815G

Only null left

Chemical Identifiers

CAS 13419-61-9
Molecular Formula C10H21NaO3S
Molecular Weight (g/mol) 244.325
MDL Number MFCD00007526
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
Synonym sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
PubChem CID 2724181
IUPAC Name sodium;decane-1-sulfonate
SMILES CCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 244.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Decanesulfonate
EINECSNumber (2)-1640
TSCA No