missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-decanesulfonate, 99%

CAS: 13419-61-9 | C10H21NaO3S | 244.325 g/mol

Supplier:  Thermo Scientific Chemicals A1463822

 View more versions of this product

Catalog No. AAA1463822


Only null left
Explore available promotions

Description

Description

Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.Sodium 1-decanesulfonate is used as a base in coupling reactions such as Suzuki coupling. It is used as an ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is used to study the mechanism of guanidine compounds on a porous graphite carbon and electro reduction of Zn2+ ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.Sodium 1-Decanesulfonate is used as a base in coupling reactions such as Suzuki coupling. It is used as an ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is used to study the mechanism of guanidine compounds on a porous graphite carbon and electro reduction of Zn2+ ions.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium 1-Decanesulfonate
>300°C
(dry wt.), Water <1.5%
MFCD00007526
3918920
Soluble in water.
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
244.325
244.33
13419-61-9
99% (dry wt.), Water <1.5%
C10H21NaO3S
100 g
sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium;decane-1-sulfonate
2724181
99%

Chemical Identifiers

13419-61-9
244.325
AIMUHNZKNFEZSN-UHFFFAOYSA-M
2724181
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
C10H21NaO3S
MFCD00007526
sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
sodium;decane-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-525-9

TSCA : No

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only