missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Butanesulfonate 98.0+%, TCI America™

Catalog No. B0379500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0379500G 500 g
B037925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0379500G Supplier TCI America Supplier No. B0379500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot
PubChem CID 4096517
IUPAC Name sodium butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 160.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Butanesulfonate