missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Butanesulfonate 98.0+%, TCI America™

Catalog No. p-7131349
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
I03425G 5 g
I034225G 25 g
2 options

Catalog No. I03425G

Supplier: TCI America I03425G

Only null left

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot
PubChem CID 4096517
IUPAC Name sodium butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 160.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Butanesulfonate [Reagent for Ion-Pair Chromatography]