missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-butanesulfonate, 99%

Catalog No. p-7020631
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1265406 5 g
AAA1265414 25 g
AAA1265422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1265406 Supplier Thermo Scientific Chemicals Supplier No. A1265406
Only null left

CAS: 2386-54-1 | C4H9NaO3S | 160.16 g/mol

Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion). Ion-associating reagent for HPLC1 and reversed-phase chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion). Ion-associating reagent for HPLC1 and reversed-phase chromatography.

Solubility
Soluble in water (1-10%).

Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot
PubChem CID 4096517
IUPAC Name sodium;butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 3716662
Solubility Information Soluble in water (1-10%).
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material Sodium 1-butanesulfonate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-201-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only