missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Sodiu mL-Lactate
Description
Sodiu mL-Lactate
Specifications
Specifications
| CAS | 867-56-1 |
| Melting Point | 163°C to 165°C |
| Molecular Formula | C3H5NaO3 |
| Linear Formula | C3H5NaO3 |
| Quantity | 50 g |
| MDL Number | MFCD00066576 |
| Synonym | (S)-2-Hydroxypropionic acid sodium salt; L-Lactic acid sodium salt; Sarcolactic acid sodium salt |
| Molecular Weight (g/mol) | 112.06 g/mol |