missing translation for 'onlineSavingsMsg'
Learn More

Soda lime, pure, granular, without indicator, Thermo Scientific™

Catalog No. AC472160010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC472160010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC472160010 Supplier Thermo Scientific Chemicals Supplier No. 472160010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.07
MDL Number 00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
IUPAC Name sodium oxocalcium hydroxide
SMILES [OH-].[Na+].O=[Ca]

Specifications

Color White to beige to light gray
Physical Form Granules
Quantity 1 kg
Loss on Drying ≤20 % (10 g, 200°C, 2 h)
Packaging Glass Bottle
Merck Index 13-8640
Solubility Information Slightly soluble in water
Grade Ungraded
Chemical Name or Material Soda lime
Hazard Category Corrosion
Recommended Storage Normal conditions