missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime, Indicating, 4-8 Mesh, Reagent, ACS, Spectrum™ Chemical

Catalog No. p-7158169
Change view
Click to view available options
Quantity:
2.5 kg
15 kg
300 lb.
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606254 300 lb. Fiber Drum
18606252 2.5 kg Poly Pail
18606253 15 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606254 Supplier Spectrum Chemical Mfg Cor Supplier No. SO102300LBBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO102, 8006-28-8

Spectrum™ Chemical Soda Lime, Indicating, 4-8 Mesh, Reagent, ACS

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.07
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
IUPAC Name sodium oxocalcium hydroxide
SMILES [OH-].[Na+].O=[Ca]

Specifications

CAS Max % 1
Quantity 300 lb.
Grade ACS
Loss on Drying 0.07
Packaging Fiber Drum