missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime, Indicating, 4-8 Mesh, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18606254
Change view
Click to view available options
Quantity:
2.5 kg
15 kg
300 lb.
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606254 300 lb. Amber Glass Bottle
18606252 2.5 kg Amber Glass Bottle
18606253 15 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606254 Supplier Spectrum Chemical Mfg Cor Supplier No. SO102300LBBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO102, 8006-28-8

Spectrum™ Chemical Soda Lime, Indicating, 4-8 Mesh, Reagent, ACS

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.07
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
IUPAC Name sodium oxocalcium hydroxide
SMILES [OH-].[Na+].O=[Ca]

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 300 lb.
Grade ACS/Reagent
Loss on Drying 0.07
Packaging Amber Glass Bottle
Chemical Name or Material Soda Lime, Indicating, 4-8 Mesh