missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime, ACS Reagent, Solstice

Catalog No. 60045719
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045719 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60045719 Supplier Solstice Supplier No. 266434500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS Reagent, granular, +100 mesh particle size

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number MFCD00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

pH 7.0 - 14.0
Color Beige
Physical Form Granular
Material Soda Lime
Quantity 500 g
Mesh Size +100mesh particle size
UN Number UN1907
Packaging Poly Bottle
Grade ACS Reagent
Chemical Name or Material Soda Lime
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H314
DOTInformation 8/PGIII