missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex Low-Profile Bottle-Top Dispensers from DWK Life Sciences™
Semiautomatic dispensers help reduce reagent waste, and accordingly, disposal costs
$1,888.75 - $3,045.93
Specifications
| Product Type | Low-profile Bottletop Dispenser |
|---|---|
| Autoclavable | Autoclavable |
| Includes | Wide mouth amber reservoir, delivery nozzle, plunger |
| Precision | <0.5 to <0.1% |
| Warranty | 1 Year |
Includes: Wide-mouth amber reservoir (also available as accessory), delivery nozzle and plunger
| Catalog Number | Mfr. No. | Volume (Metric) | Increments | Volume (English) Reservoir | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Increments | Volume (English) Reservoir | Price | Quantity & Availability | |||
|
1370618
|
Socorex
844068 |
1 to 30 mL | 1 mL | 67.62 oz. |
Each for $3,045.93
|
|
|||
|
1370617
|
Socorex
844064 |
1 to 10 mL | 0.2 mL | 33.81 oz. |
Each for $2,382.21
|
|
|||
|
1370616
|
Socorex
844060 |
0.2 to 2 mL | 0.1 mL | 8.45 oz. |
Each for $1,888.75
|
|
|||
Description
- Retractable, graduated column makes volume setting easy and saves space in incubators and refrigerators
- PTFE resin coated plunger prevents the “freezing” normally associated with dispensing caustic solutions
- Delivery nozzle locks into a threaded fitting for safe operation
- No metal parts or springs to corrode; all parts that come in contact with the liquid are resistant to most chemicals
- Valve is made of borosilicate glass; valve balls are of synthetic ruby
- Syringe is made of borosilicate glass; delivery jet and unions are made of PTFE resin
- Three models cover volume range: 0.2 to 30mL
Order Info
Can be removed for cleaning and/or autoclaving (121°C/250°F)
Specifications
| Low-profile Bottletop Dispenser | |
| Wide mouth amber reservoir, delivery nozzle, plunger | |
| 1 Year |
| Autoclavable | |
| <0.5 to <0.1% |
Not for use with concentrated or full-strength acids