missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soap Solution, 1 mL = 1 mg CaCO3, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Soap Solution |
| Color | Colorless |
| CAS | 64-17-5, 7732-18-5, 8029-38-7 |
| Physical Form | Liquid |
| CAS Min % | 74.0, 23.3, 1.17 |
| CAS Max % | 77.0, 24.25, 1.21 |
| Quantity | 1 L |
| Packaging | Natural Poly Bottle |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 1 mL = 1 mg CaCO3 |