missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sneeze-Guard, Upright Mounted

Catalog No. 25100627
Change view
Click to view available options
Height (English):
14 in.
26 in.
38 in.
Width (English):
48 in.
60 in.
72 in.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English)
25100627 26 in. 72 in.
25100620 14 in. 48 in.
25100623 14 in. 60 in.
25100626 14 in. 72 in.
25100621 26 in. 48 in.
25100624 26 in. 60 in.
25100622 38 in. 48 in.
25100625 38 in. 60 in.
25100628 38 in. 72 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 25100627 Supplier Fisherbrand™ Supplier No. GPU2672/PEND
Only null left

Get protected while you are working with this Fisherbrand™ Sneeze Guard Panel. It is easy to setup and easy to clean.

Clean with soap and water. Antibacterial sprays are ok to use as well.

Specifications

Width (English) 72 in.
Height (English) 26 in.
Lens Tint Clear
Product Type Sneeze Guard