missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sneeze-Guard, Edge Mounted

Catalog No. 25100632
Change view
Click to view available options
Height (English):
18 in.
24 in.
Width (English):
24 in.
30 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English)
25100632 24 in. 30 in.
25100629 18 in. 24 in.
25100631 18 in. 30 in.
25100630 24 in. 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 25100632 Supplier Fisherbrand™ Supplier No. GPE2430/PEND

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Get protected while you are working with this Fisherbrand™ Sneeze Guard Panel. It is easy to setup and easy to clean.

Clean with soap and water. Antibacterial sprays are ok to use as well.

Specifications

Width (English) 30 in.
Height (English) 24 in.
Lens Tint Clear
Product Type Sneeze Guard