Learn More
New England Biolab Ltd SNAP-Surface Alexa Fluor 647 50 nmol
Supplier: New England Biolab Ltd S9136S
SNAP-Surface Alexa Fluor 647 is a photostable red fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.