missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SNAP i.d.™ Starter Package, Mini and Midi, 7.5cm and 8.5 x 13.5cm

Catalog No. STARTMM
Change view
Click to view available options
Product Type:
Western Blot Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
STARTMM Western Blot Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STARTMM Supplier MilliporeSigma™ Supplier No. STARTMM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • SNAP i.d.™ 2.0 Protein Detection System
  • Immobilon™-P Membrane, 0.45μm, 26.5cm x 3.75m roll
  • SNAP i.d.™ 2.0 Blot Holders, 100 units/pk
  • Bløk™-CH Noise Cancelling Reagent
  • Luminata™ Classico Western HRP Substrate, 100mL
  • Reblot™ Plus Strong Antibody Stripping Solution, 10X, 50mL

Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs! The SNAP i.d.™ 2.0 system applies a vacuum to actively drive reagents through the membrane, promoting antigen binding and thorough washing, enabling you to better optimize your Western blotting.

  • Superior: Increased antibody-antigen binding, enhanced washes, and antibody recollection
  • Flexible: Two gel sizes, mini (7.5 x 8.4cm) and midi (8.7 x 13.5cm)
  • Fast: 30-minute immunodetection
  • Faster testing of different antibodies
  • Higher throughput of Western blots each day
  • Draws reagents through blotting membrane
  • Minimizes over-blocking
  • Thoroughly flushes membranes instead of just rinsing
  • Reduces incubation times

Specifications

Product Type Western Blot Kit
For Use With (Application) Western Blot