missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SNAP i.d.™ Starter Package, Midi, 8.5 x 13.5cm

Catalog No. STARTMIDI
Change view
Click to view available options
Quantity:
1 Midi Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STARTMIDI 1 Midi Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STARTMIDI Supplier MilliporeSigma™ Supplier No. STARTMIDI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • SNAP i.d.™ 2.0 Protein Detection System
  • Immobilon™-P Membrane, 0.45μm, 26.5cm x 3.75m roll
  • SNAP i.d.™ 2.0 Blot Holders, 100 units/pk
  • Bløk™-CH Noise Cancelling Reagent
  • Luminata™ Classico Western HRP Substrate, 100mL
  • Reblot™ Plus Strong Antibody Stripping Solution, 10X, 50mL

Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs! The SNAP i.d.™ 2.0 system applies a vacuum to actively drive reagents through the membrane, promoting antigen binding and thorough washing, enabling you to better optimize your Western blotting.

  • Superior: Increased antibody-antigen binding, enhanced washes, and antibody recollection
  • Flexible: Two gel sizes, mini (7.5 x 8.4cm) and midi (8.7 x 13.5cm)
  • Fast: 30-minute immunodetection
  • Faster testing of different antibodies
  • Higher throughput of Western blots each day
  • Draws reagents through blotting membrane
  • Minimizes over-blocking
  • Thoroughly flushes membranes instead of just rinsing
  • Reduces incubation times

Specifications

Product Type Western Blot Kit
For Use With (Application) Western Blot
Quantity 1 Midi Kit