Learn More
Brady™ Snap Cap Gas Cylinder Lockout
Catalog No. 19103972
Change view
Click to view available options
Product Type:
High Pressure, Coarse Thread Cylinder Lockout
High Pressure, Fine Thread Cylinder Lockout
Low Pressure, Coarse Thread Cylinder Lockout
Low Pressure, Fine Thread Cylinder Lockout
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19103972 Low Pressure, Coarse Thread Cylinder Lockout
19103969 High Pressure, Fine Thread Cylinder Lockout
19103970 Low Pressure, Fine Thread Cylinder Lockout
19103971 High Pressure, Coarse Thread Cylinder Lockout
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19103972 Supplier Brady™ Supplier No. 95139
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deny access to the main cylinder control and regulator attachment of gas cylinders. The Brady™ Snap Gas Cylinder Lockout fits most North American high and low pressure gas cylinders and is made to last many years of use.
  • Designed to fit most North American high and low pressure gas cylinders
  • Snap Cap devices denies access to main cylinder control and regulator attachment
  • Made of weld steel and powder coated for many years of use
  • Operates safely from -31° F to 203° F
Specifications
Product Type Low Pressure, Coarse Thread Cylinder Lockout
Color Yellow
Material Steel
Description Thread Type: Coarse
Dimensions (L x W x H) 6.5 x 3 1/8 x 11 UN
Finish Powder-coated
Quantity 1 Ea.