missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SNAP, 98%

Catalog No. AAJ67204FPL
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67204FPL 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67204FPL Supplier Thermo Scientific Chemicals Supplier No. J67204FPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79032-48-7 | C7H12N2O4S, C7H12N2O4S | 220.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79032-48-7
Molecular Formula C7H12N2O4S
Synonym S-Nitroso-N-acetyl-D,L-penicillamine;

Specifications

Quantity 20 mg
Description Green
Content And Storage -30°C to -10°C
Formula Weight 220.25
Solubility Soluble in DMSO at 20mg/ml
Product Type SNAP, 98%
Form Crystalline solid
Assay 98%

RUO – Research Use Only