missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ SMS Pleated Bouffant Caps

Catalog No. 19472796
Catalog No. 19472796 Supplier International Enviroguard™ Supplier No. 210NW21PL

Pleated bouffants are lightweight, breathable, and provide an secure particulate barrier. Three-layer SMS fabric keeps hair in place, while also keeping worker's cool for extended periods.

  • SMS pleated bouffants are lightweight and breathable
  • Durable, three-layer SMS fabric keeps hair securely in place
  • Protects hair from contaminants like dust and other particles, while also protecting products from cross-contamination
  • Expandable fit with an elastic closure comfortably accommodates a wide range of hairstyles without pulling when removed
  • Potential applications include healthcare, pharmaceutical, labs, critical environments, food processing, and more

Specifications

Product Type Cleanroom Bouffant
Material SMS Fabric
Color White
Disposable Single-use
Latex Free Yes
Model 210NW
For Use With (Application) Healthcare, Pharma, Labs, Critical Environments, Food Processing
Style Pleated
Cleanroom Class Class 100 (ISO 5)