missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety White SMS Lab Coats, Snap Front, Knit Wrists

Catalog No. 19168632
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19168632 X-Large
19055316 Small
19168630 Medium
19168631 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19168632 Supplier Keystone Safety Supplier No. LC3WKSMSXLWHITE

missing translation for 'validMainframeMsg'

Latex free SMS lab coats with three convenient pockets offer barrier protection while keeping the wearer cool and comfortable.

  • Latex free, 3-layer SMS fabric
  • Barrier against water based liquids and light chemical splash
  • Single collar
  • Strong, high tensile strength
  • Comfortable, lightweight, breathable
  • Serged seams
  • Meets ANSI 101-1996
  • Multi use for cleanroom, pharmaceutical, food, janitorial, cleaning, maintenance
  • Keystone LC3-WK-SMS-WHITE SMS Lab Coat, 3 Pockets, Knit Wrists, Snap Front
  • Pack of 30

Specifications

Product Type Controlled Environments Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Collar Type Single
Latex Free Yes
Model LC3-WK-SMS
Seam Type Serged
Certifications/Compliance ANSI 101-1996
For Use With (Application) Janitorial, Manufacturing, Food Service, Industrial, Medical, Cleanroom, Pharmaceutical
Length (English) 40 in.
Length (Metric) 101.6 cm
Weight (English) 1.4 oz.
No. per Pack 30/Pk.
Show More Show Less