missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety White SMS Lab Coats, Snap Front, Knit Wrists

Catalog No. 19055316
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19055316 Small
19168630 Medium
19168631 Large
19168632 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19055316 Supplier Keystone Safety Supplier No. LC3WKSMSSMWHITE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Latex free SMS lab coats with three convenient pockets offer barrier protection while keeping the wearer cool and comfortable.

  • Latex free, 3-layer SMS fabric
  • Barrier against water based liquids and light chemical splash
  • Single collar
  • Strong, high tensile strength
  • Comfortable, lightweight, breathable
  • Serged seams
  • Meets ANSI 101-1996
  • Multi use for cleanroom, pharmaceutical, food, janitorial, cleaning, maintenance
  • Keystone LC3-WK-SMS-WHITE SMS Lab Coat, 3 Pockets, Knit Wrists, Snap Front
  • Pack of 30

Specifications

Product Type Controlled Environments Lab Coat
Size Small
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Collar Type Single
Latex Free Yes
Model LC3-WK-SMS
Seam Type Serged
Certifications/Compliance ANSI 101-1996
For Use With (Application) Janitorial, Manufacturing, Food Service, Industrial, Medical, Cleanroom, Pharmaceutical
Autoclavable Not Autoclavable
No. per Pack 30/Pk.
Show More Show Less