missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety White SMS Disposable Anti-Static Frocks

Catalog No. 19055482
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19055482 3X-Large
19055476 Small
19055478 Large
19055479 X-Large
19055480 2X-Large
19055483 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19055482 Supplier Keystone Safety Supplier No. CEF3WKSMS3XLWHITE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Frocks constructed of SMS composite fabric feature knit wrists, elastic thumb loops, and three pockets. They provide particulate and liquid barrier suitable for ISO Class 6 cleanrooms.

  • Made from SMS composite fabric
  • Disposable
  • Highly breathable and comfortable
  • ISO Class 6 cleanroom compatible
  • Low shedding and anti-static to protect sensitive products
  • Provide particulate barrier and liquid repellant
  • Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
  • Knit cuffs with elastic thumb loops, three pockets, six snap closure, knit collar, extra-long knee-length, hanger loop

Specifications

Size 3X-Large
Material SMS
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Knit, Elastic Thumb Loop
Collar Type Knit
Latex Free Yes
No. of Pockets 3
Sleeve Style Long Sleeve
Packaging Type 10/Bag, 3 Bags/Lined Carton
Cleanroom Class Class 1000 (ISO 6)
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less