missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Safety White SMS Disposable Anti-Static Frocks
Frocks constructed of SMS composite fabric feature knit wrists, elastic thumb loops, and three pockets. They provide particulate and liquid barrier suitable for ISO Class 6 cleanrooms.
Supplier: Keystone Safety CEF3WKSMS2XLWHITE
Description
- Made from SMS composite fabric
- Disposable
- Highly breathable and comfortable
- ISO Class 6 cleanroom compatible
- Low shedding and anti-static to protect sensitive products
- Provide particulate barrier and liquid repellant
- Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
- Knit cuffs with elastic thumb loops, three pockets, six snap closure, knit collar, extra-long knee-length, hanger loop
Spécifications
| Controlled Environments Frock | |
| SMS | |
| Single-use | |
| Snaps | |
| Serged | |
| Knit | |
| 3 | |
| 10/Bag, 3 Bags/Lined Carton |
| 2X-Large | |
| White | |
| Non-sterile | |
| Knit, Elastic Thumb Loop | |
| Manufacturing, Food Service, Industrial, Medical, Pharmaceutical | |
| Yes | |
| Long Sleeve | |
| Class 1000 (ISO 6) |