missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ IBA-35, X-Heavy Chemical Resistant Butyl Gloves
Extra heavyweight butyl gloves are 35 mil thick and feature a smooth finish to protect against a wide range of hazardous chemicals.
Supplier: Guardian™ 53504
Description
- Extra heavy butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 35 mil thickness, 14 in. length
- Made in the USA
Specifications
| ASTM F739-05 | |
| Black | |
| Yes | |
| Powdered | |
| 35 mil | |
| Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones | |
| Smooth | |
| ISO 9001 | |
| 14 in. |
| CE | |
| Specific | |
| Butyl | |
| Large | |
| 0.88 mm | |
| Gauntlet/Rolled | |
| Unlined | |
| 53504 | |
| 35.5 cm |