missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SMC4 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 10051 |
| Species | Human |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56394 |
| Gene Alias | CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog |
| Gene Symbol | SMC4 |
| Quantity | 0.1 mg |
| Research Category | Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers |
| Show More |