Alkali Scientific SmartReader™ 96 Microplate Absorbance Reader, for 96 Well Plates, 115V QTY 1
Catalog No. 502336120
The Accuris SmartReader 96 is a welcome addition to any lab that is routinely measuring concentration or absorbance in 96 well plates. The instruments 7 inch touch screen, intuitive software and graphical interface makes it easy to use as a standalone instrument (a separate computer and additional software is not required).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Accuris SmartReader 96 is a welcome addition to any lab that is routinely measuring concentration or absorbance in 96 well plates. The instruments 7 inch touch screen, intuitive software and graphical interface makes it easy to use as a standalone instrument (a separate computer and additional software is not required).