missing translation for 'onlineSavingsMsg'
Learn More

Wanzl SmartCell™ Shelving 3-Position Label Holder

Catalog No. 11222165
Change view
Click to view available options
Color:
Blue
Clear
Green
Orange
Pink
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11222165 Clear
11222164 Blue
11222166 Green
11222167 Orange
11222168 Pink
11222169 Red
11222170 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 11222165 Supplier Wanzl Supplier No. T.LR03CL
Only null left

Label Holder for SmartCell™ Shelving.

  • Accessory for SmartCell™ Shelving
  • 3 in. 3-Position Label Holders
  • 50/Bag

Specifications

Includes 50 Label Holders
Length (English) 3 in.
With Casters No
Color Clear
For Use With (Application) Shelving
Material Plastic