missing translation for 'onlineSavingsMsg'
Learn More

Wanzl SmartCell™ Post Mount Divider

Catalog No. 11222066
Change view
Click to view available options
Width (English):
10 in.
18 in.
24 in.
46 in.
48 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11222066 10 in.
11222062 18 in.
11222063 24 in.
11222064 46 in.
11222065 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11222066 Supplier Wanzl Supplier No. T.HPM410DV

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Post mount dividers allow for easy custom configuration of mobile carts for greater mobility and visibility.

  • Allow for custom compartmentalization of the SmartCell™ basket system.
  • Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval.

Specifications

Color Chrome
Material Steel
For Use With (Application) Shelving
Shelf Type Divider
Width (English) 10 in.