missing translation for 'onlineSavingsMsg'
Learn More

Wanzl SmartCell™ Clear Wire Shelf Label Holder

Catalog No. p-7224991
Change view
Click to view available options
Width (English):
18 in.
36 in.
48 in.
60 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11222031 18 in.
11222032 36 in.
11222033 48 in.
11222077 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11222031 Supplier Wanzl Supplier No. T.LHB18CL
Only null left

Wire label holder for SmartCell™ Shelving System.

  • Add-on option for additional customization
  • Clear
  • Three ways to mount

Specifications

Color Chrome
Material Steel
For Use With (Application) Shelving
For Use With (Equipment) SmartCell™
Shelf Type Basket Label Holder
Width (English) 18 in.