missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl SmartCell™ Chrome Shelving Divider
Description
- Allow for custom compartmentalization of the SmartCell™ basket system.
- Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval
Specifications
Specifications
| Color | Chrome |
| Material | Steel |
| Dimensions (L x W) | 18 x 6 in. |
| For Use With (Application) | Shelving |
| For Use With (Equipment) | SmartCell™ |
| Includes | 1 Divider |
| Width (English) | 6 in. |