missing translation for 'onlineSavingsMsg'
Learn More

Wanzl SmartCell™ Chrome Shelving Divider

Catalog No. 11222027
Change view
Click to view available options
Dimensions (L x W):
3 x 6 in.
6 x 6 in.
12 x 6 in.
18 x 6 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
11222027 3 x 6 in.
11222028 6 x 6 in.
11222029 12 x 6 in.
11222030 18 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11222027 Supplier Wanzl Supplier No. T.H36DV
Only null left

Chrome Plus Basket Divider for SmartCell Shelving System.

  • Allow for custom compartmentalization of the SmartCell™ basket system.
  • Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval

Specifications

Color Chrome
Material Steel
Dimensions (L x W) 3 x 6 in.
For Use With (Application) Shelving
For Use With (Equipment) SmartCell™
Includes 1 Divider
Width (English) 6 in.