Learn More
Thermo Scientific™ Smart-Vue™ Repeater Modules
Extend the monitoring distance of Thermo Scientific™ Smart-Vue™ Wireless Monitoring Solutions by using these repeater modules.
Supplier:  Thermo Scientific™ SV100500LSB
 View more versions of this product
Catalog No. SV100500LSB
Only null left
Description
Description
Extend the monitoring distance of Thermo Scientific™ Smart-Vue™ Wireless Monitoring Solutions by using Thermo Scientific™ Smart-Vue™ Repeater Modules.
Includes:
Repeater module, universal (100/240V AC) power supply, wall-mounting holder w/velcro and customer reference letter.
Specifications
Specifications
Repeater Module
Not Calibrated
Smart-Vue 25/500mW Repeater Module (915MHz)
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Smart-Vue™ Wireless Monitoring Solutions
3000 Readings
Smart-Vue Client, Smart-Vue Server
25/500mW Repeater module, universal (100/240V AC) power supply, wall-mounting holder w/velcro, and customer reference letter
Repeater Module
Product Suggestions
Product Suggestions