Learn More
Thermo Scientific™ Smart-Vue Pro Cloud Software and Subscription Services
Catalog No. SVSWCS22000
Change view
Click to view available options
For Use With (Application):
Up to 100 sensors
Up to 1000 sensors
Up to 1500 sensors
Up to 20 sensors
Up to 2000 sensors
Up to 250 sensors
Up to 5 sensors
Up to 50 sensors
Up to 500 sensors
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
SVSWCS22000 Up to 2000 sensors
SVSWCS20005 Up to 5 sensors
SVSWCS20020 Up to 20 sensors
SVSWCS20050 Up to 50 sensors
SVSWCS20100 Up to 100 sensors
SVSWCS20250 Up to 250 sensors
SVSWCS20500 Up to 500 sensors
SVSWCS21000 Up to 1000 sensors
SVSWCS21500 Up to 1500 sensors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. SVSWCS22000 Supplier Thermo Scientific™ Supplier No. SVPSWCS22000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times.
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud or customer server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • And trail and secure-level access control to support 21CFR Part 11
  • Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Specifications
Type Smart-Vue Pro Software and Subscription Service
Description Smart-Vue Pro Cloud license key - Access to the Cloud and Smart-Vue Pro Web application for installation up to 2000 sensors. License validity: 12 months after activation
Certifications/Compliance Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
For Use With (Application) Up to 2000 sensors
For Use With (Equipment) Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module