Learn More
Thermo Scientific™ Smart-Vue Pro Alert Software
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions, Smart-Vue™ Alert will automatically notify the user based notification preferences wherever you are (phone, text or email).
- Intuitive dashboard simplifies daily usage.
- Optional floor plan and sites views.
- Data can be exported with a click for reporting and customizable graphs.
- Web application on Cloud or customer server.
- Instant access to alarms, readings, and graphs.
- Remote data access.
- Flexible organization by site, department user, equipment, and criticality.
- And trail and secure-level access control to support 21CFR Part 11
- Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Specifications
Specifications
| Type | Smart-Vue Pro Software and Subscription Service |
| Description | Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 50 PTS |
| Certifications/Compliance | Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance |
| For Use With (Application) | Up to 50 PTS |
| For Use With (Equipment) | Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |