Learn More
Thermo Scientific™ Smart-Vue Pro Alert Software
Catalog No. SVSWSA20005
Change view
Click to view available options
For Use With (Application):
Up to 100 PTS
Up to 1000 PTS
Up to 1500 PTS
Up to 20 PTS
Up to 250 PTS
Up to 2500 PTS
Up to 5 PTS
Up to 50 PTS
Up to 500 PTS
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
SVSWSA20005 Up to 5 PTS
SVSWSA20020 Up to 20 PTS
SVSWSA20050 Up to 50 PTS
SVSWSA20100 Up to 100 PTS
SVSWSA20250 Up to 250 PTS
SVSWSA20500 Up to 500 PTS
SVSWSA21000 Up to 1000 PTS
SVSWSA21500 Up to 1500 PTS
SVSWSA22000 Up to 2500 PTS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. SVSWSA20005 Supplier Thermo Scientific™ Supplier No. SVPSWSA20005
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions, Smart-Vue™ Alert will automatically notify the user based notification preferences wherever you are (phone, text or email).
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud or customer server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • And trail and secure-level access control to support 21CFR Part 11
  • Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Specifications
Type Smart-Vue Pro Software and Subscription Service
Description Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 5 PTS
For Use With (Application) Up to 5 PTS
For Use With (Equipment) Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module